Assay Details
| Gene Symbol: Lins | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499399.2 | XP_003499447.1 | 4 - 5 | 696 | 90 | |
| XM_007640152.1 | XP_007638342.1 | 4 - 5 | 787 | 90 | ||
| XM_007640153.1 | XP_007638343.1 | 4 - 5 | 746 | 90 | ||
| GenBank mRNA | JI882981.1 | - | 4 - 5 | 718 | 90 | |
| JP039043.1 | - | 4 - 5 | 718 | 90 | ||

Lins
Genomic Map