Assay Details
| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653684.1 | XP_007651874.1 | 3 - 4 | 643 | 69 | |
| GenBank mRNA | JP058259.1 | - | 3 - 4 | 636 | 69 | |
More Information
Set Membership: |
|

Tmem41b
Genomic Map