Assay Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503249.2 | XP_003503297.1 | 3 - 4 | 394 | 60 | |
| GenBank mRNA | JI866365.1 | - | 3 - 4 | 184 | 60 | |
More Information
Set Membership: |
|

Ppif
Genomic Map