Assay Details
| Gene Symbol: Lpar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506970.2 | XP_003507018.1 | 3 - 4 | 968 | 66 | |
| XM_007647502.1 | XP_007645692.1 | 4 - 5 | 1083 | 66 | ||
| XM_007647503.1 | XP_007645693.1 | 3 - 4 | 940 | 66 | ||
| XM_007647504.1 | XP_007645694.1 | 3 - 4 | 959 | 66 | ||
| GenBank mRNA | JI889075.1 | - | 1 - 2 | 158 | 66 | |
| JP065951.1 | - | 1 - 2 | 158 | 66 | ||
More Information
Set Membership: |
|

Lpar1
Genomic Map