Assay Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514217.2 | XP_003514265.1 | 2 - 3 | 322 | 78 | |
| GenBank mRNA | JI871242.1 | - | 1 - 2 | 283 | 78 | |
More Information
Set Membership: |
|

Ngrn
Genomic Map