Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645927.1 | XP_007644117.1 | 3 - 4 | 527 | 87 | |
| GenBank mRNA | JI896500.1 | - | 3 - 4 | 528 | 87 | |
More Information
Set Membership: |
|

Osgin2
Genomic Map