Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654533.1 | XP_007652723.1 | 3 - 4 | 458 | 79 | |
| GenBank mRNA | JI875099.1 | - | 3 - 4 | 458 | 79 | |
More Information
Set Membership: |
|

Gatad1
Genomic Map