Assay Details
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007608908.1 | XP_007607098.1 | 2 - 3 | 263 | 60 | |
GenBank mRNA | JI874616.1 | - | 2 - 3 | 243 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
