Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654408.1 | XP_007652598.1 | 12 - 13 | 2194 | 58 | |
| XM_007654409.1 | XP_007652599.1 | 12 - 13 | 1920 | 58 | ||
| GenBank mRNA | JI865402.1 | - | 12 - 13 | 1683 | 58 | |
More Information
Set Membership: |
|

Fam149a
Genomic Map