Assay Details
| Gene Symbol: Znf329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650127.1 | XP_007648317.1 | 1 - 2 | 704 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf329| Gene Symbol: Znf329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650127.1 | XP_007648317.1 | 1 - 2 | 704 | 118 | |
Set Membership: |
|