Assay Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496646.2 | XP_003496694.1 | 4 - 5 | 962 | 74 | |
| GenBank mRNA | JI881085.1 | - | 4 - 5 | 946 | 74 | |
| JP057399.1 | - | 4 - 5 | 946 | 74 | ||
More Information
Set Membership: |
|

Medag
Genomic Map