Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510114.2 | XP_003510162.1 | 4 - 5 | 1801 | 63 | |
| XM_007650750.1 | XP_007648940.1 | 5 - 6 | 836 | 63 | ||
| XM_007650751.1 | XP_007648941.1 | 5 - 6 | 1071 | 63 | ||
| XM_007650752.1 | XP_007648942.1 | 5 - 6 | 1069 | 63 | ||
| GenBank mRNA | JI862791.1 | - | 4 - 5 | 1787 | 63 | |
| JP042920.1 | - | 4 - 5 | 1787 | 63 | ||
More Information
Set Membership: |
|

Tmem219
Genomic Map