Assay Details
Gene Symbol: Sstr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511009.2 | XP_003511057.1 | 1 - 2 | 184 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
