Assay Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653496.1 | XP_007651686.1 | 2 - 3 | 450 | 64 | |
| GenBank mRNA | JI863276.1 | - | 2 - 3 | 282 | 64 | |
| JP065338.1 | - | 2 - 3 | 282 | 64 | ||
| M99692.1 | - | 2 - 3 | 422 | 64 | ||
More Information
Set Membership: |
|

Fth1
Genomic Map