Assay Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497563.2 | XP_003497611.1 | 4 - 5 | 402 | 65 | |
| XM_007613978.1 | XP_007612168.1 | 4 - 5 | 419 | 65 | ||
| GenBank mRNA | JI870678.1 | - | 1 - 2 | 70 | 65 | |
More Information
Set Membership: |
|

Dnajc24
Genomic Map