Assay Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655205.1 | XP_007653395.1 | 9 - 10 | 1650 | 65 | |
GenBank mRNA | JI894477.1 | - | 6 - 7 | 1066 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
