Assay Details
| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649074.1 | XP_007647264.1 | 4 - 5 | 715 | 69 | |
| GenBank mRNA | JI878737.1 | - | 1 - 2 | 135 | 69 | |
More Information
Set Membership: |
|

Hif1an
Genomic Map