Assay Details
Gene Symbol: Nhlrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646092.1 | XP_007644282.1 | 1 | 124 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
