Assay Details
| Gene Symbol: Polr3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496464.2 | XP_003496512.1 | 8 - 9 | 954 | 73 | |
| GenBank mRNA | JP065182.1 | - | 8 - 9 | 952 | 73 | |
More Information
Set Membership: |
|

Polr3f
Genomic Map