Assay Details
| Gene Symbol: Spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650156.1 | XP_007648346.1 | 3 - 4 | 496 | 105 | |
| GenBank mRNA | JP058409.1 | - | 3 - 4 | 493 | 105 | |
More Information
Set Membership: |
|

Spcs3
Genomic Map