Assay Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500145.2 | XP_003500193.2 | 2 - 3 | 236 | 83 | |
| GenBank mRNA | JI878124.1 | - | 2 - 3 | 236 | 83 | |
| JP059251.1 | - | 2 - 3 | 236 | 83 | ||
More Information
Set Membership: |
|

Desi2
Genomic Map