Assay Details
| Gene Symbol: Mrps33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643649.1 | XP_007641839.1 | 2 - 3 | 341 | 65 | |
| GenBank mRNA | JI881021.1 | - | 2 - 3 | 323 | 65 | |
More Information
Set Membership: |
|

Mrps33
Genomic Map