Assay Details
| Gene Symbol: Tgm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007637407.1 | XP_007635597.1 | 7 - 8 | 1070 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgm6| Gene Symbol: Tgm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007637407.1 | XP_007635597.1 | 7 - 8 | 1070 | 62 | |
Set Membership: |
|