Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654535.1 | XP_007652725.1 | 4 - 5 | 754 | 76 | |
| XM_007654536.1 | XP_007652726.1 | 4 - 5 | 752 | 76 | ||
| XM_007654539.1 | XP_007652729.1 | 3 - 4 | 370 | 76 | ||
| XM_007654540.1 | XP_007652730.1 | 3 - 4 | 483 | 76 | ||
| XM_007654541.1 | XP_007652731.1 | 3 - 4 | 456 | 76 | ||
| XM_007654542.1 | XP_007652732.1 | 3 - 4 | 329 | 76 | ||
| XM_007654543.1 | XP_007652733.1 | 3 - 4 | 476 | 76 | ||
More Information
Set Membership: |
|

LOC100770307
Genomic Map