Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654535.1 | XP_007652725.1 | 6 - 7 | 919 | 102 | |
| XM_007654536.1 | XP_007652726.1 | 6 - 7 | 917 | 102 | ||
| XM_007654537.1 | XP_007652727.1 | 5 - 6 | 865 | 102 | ||
| XM_007654538.1 | XP_007652728.1 | 5 - 6 | 862 | 102 | ||
| XM_007654539.1 | XP_007652729.1 | 5 - 6 | 535 | 102 | ||
| XM_007654540.1 | XP_007652730.1 | 5 - 6 | 648 | 102 | ||
| XM_007654541.1 | XP_007652731.1 | 5 - 6 | 621 | 102 | ||
| XM_007654542.1 | XP_007652732.1 | 5 - 6 | 494 | 102 | ||
| XM_007654543.1 | XP_007652733.1 | 5 - 6 | 641 | 102 | ||
| XM_007654544.1 | XP_007652734.1 | 4 - 5 | 470 | 102 | ||
More Information
Set Membership: |
|

LOC100770307
Genomic Map