Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654535.1 | XP_007652725.1 | 7 - 8 | 1021 | 98 | |
| XM_007654536.1 | XP_007652726.1 | 7 - 8 | 1019 | 98 | ||
| XM_007654537.1 | XP_007652727.1 | 6 - 7 | 967 | 98 | ||
| XM_007654538.1 | XP_007652728.1 | 6 - 7 | 964 | 98 | ||
| XM_007654539.1 | XP_007652729.1 | 6 - 7 | 637 | 98 | ||
| XM_007654540.1 | XP_007652730.1 | 6 - 7 | 750 | 98 | ||
| XM_007654541.1 | XP_007652731.1 | 6 - 7 | 723 | 98 | ||
| XM_007654542.1 | XP_007652732.1 | 6 - 7 | 596 | 98 | ||
| XM_007654543.1 | XP_007652733.1 | 6 - 7 | 743 | 98 | ||
| XM_007654544.1 | XP_007652734.1 | 5 - 6 | 572 | 98 | ||
More Information
Set Membership: |
|

LOC100770307
Genomic Map