Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654535.1 | XP_007652725.1 | 8 - 9 | 1199 | 85 | |
| XM_007654537.1 | XP_007652727.1 | 7 - 8 | 1145 | 85 | ||
| XM_007654539.1 | XP_007652729.1 | 7 - 8 | 815 | 85 | ||
| XM_007654540.1 | XP_007652730.1 | 7 - 8 | 928 | 85 | ||
| XM_007654541.1 | XP_007652731.1 | 7 - 8 | 901 | 85 | ||
| XM_007654542.1 | XP_007652732.1 | 7 - 8 | 774 | 85 | ||
| XM_007654543.1 | XP_007652733.1 | 7 - 8 | 921 | 85 | ||
More Information
Set Membership: |
|

LOC100770307
Genomic Map