Assay Details
| Gene Symbol: Mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515322.2 | XP_003515370.1 | 2 - 3 | 143 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mb| Gene Symbol: Mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515322.2 | XP_003515370.1 | 2 - 3 | 143 | 75 | |