Assay Details
Gene Symbol: Wdr83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501937.2 | XP_003501985.1 | 3 - 4 | 533 | 71 | |
GenBank mRNA | JI888342.1 | - | 3 - 4 | 517 | 71 | |
JP048220.1 | - | 3 - 4 | 517 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
