Assay Details
| Gene Symbol: Sacs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641257.1 | XP_007639447.1 | 6 - 7 | 2021 | 131 | |
| XM_007641258.1 | XP_007639448.1 | 2 - 3 | 244 | 131 | ||
| GenBank mRNA | JI863785.1 | - | 2 - 3 | 271 | 131 | |
| JP063157.1 | - | 2 - 3 | 271 | 131 | ||
More Information
Set Membership: |
|

Sacs
Genomic Map