Assay Details
| Gene Symbol: Cldn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502508.1 | XP_003502556.1 | 4 - 5 | 615 | 55 | |
| XM_007643130.1 | XP_007641320.1 | 4 - 5 | 615 | 55 | ||
More Information
Set Membership: |
|

Cldn18
Genomic Map