Assay Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640957.1 | XP_007639147.1 | 3 - 4 | 472 | 74 | |
| GenBank mRNA | JP040944.1 | - | 3 - 4 | 482 | 74 | |
More Information
Set Membership: |
|

Fbxo28
Genomic Map