Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495672.2 | XP_003495720.1 | 8 - 9 | 791 | 58 | |
| XM_007643124.1 | XP_007641314.1 | 8 - 9 | 788 | 58 | ||
| GenBank mRNA | JI861916.1 | - | 6 - 7 | 601 | 58 | |
More Information
Set Membership: |
|

Tmem104
Genomic Map