Assay Details
Gene Symbol: Cnot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499807.2 | XP_003499855.1 | 3 - 4 | 902 | 61 | |
GenBank mRNA | JI863714.1 | - | 3 - 4 | 361 | 61 | |
JP059674.1 | - | 3 - 4 | 361 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
