Assay Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643637.1 | XP_007641827.1 | 2 - 3 | 333 | 65 | |
| GenBank mRNA | JP059020.1 | - | 1 | 407 | 65 | |
More Information
Set Membership: |
|

Mospd1
Genomic Map