Assay Details
| Gene Symbol: Pcdhga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508202.1 | XP_003508250.1 | 1 | 1425 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga6| Gene Symbol: Pcdhga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508202.1 | XP_003508250.1 | 1 | 1425 | 76 | |
Set Membership: |
|