Assay Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505582.2 | XP_003505630.1 | 7 - 8 | 695 | 73 | |
| GenBank mRNA | JI870628.1 | - | 2 - 3 | 213 | 73 | |
More Information
Set Membership: |
|

Lin37
Genomic Map