Assay Details
Gene Symbol: Cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506479.2 | XP_003506527.1 | 4 - 5 | 411 | 71 | |
GenBank mRNA | JI877093.1 | - | 3 - 4 | 347 | 71 | |
JP039739.1 | - | 3 - 4 | 349 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
