Assay Details
| Gene Symbol: Rrp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494975.2 | XP_003495023.1 | 3 - 4 | 556 | 70 | |
| GenBank mRNA | JP064556.1 | - | 3 - 4 | 587 | 70 | |
More Information
Set Membership: |
|

Rrp15
Genomic Map