Assay Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654268.1 | XP_007652458.1 | 4 - 5 | 861 | 64 | |
| XM_007654269.1 | XP_007652459.1 | 4 - 5 | 861 | 64 | ||
More Information
Set Membership: |
|

C8g
Genomic Map