Assay Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515376.2 | XP_003515424.1 | 4 - 5 | 448 | 68 | |
| XM_007607580.1 | XP_007605770.1 | 4 - 5 | 448 | 68 | ||
| GenBank mRNA | JI887113.1 | - | 4 - 5 | 447 | 68 | |
| JP051167.1 | - | 4 - 5 | 447 | 68 | ||
More Information
Set Membership: |
|

Adh7
Genomic Map