Assay Details
| Gene Symbol: Sec61g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642992.1 | XP_007641182.1 | 1 - 2 | 258 | 79 | |
| XM_007642993.1 | XP_007641183.1 | 1 - 2 | 258 | 79 | ||
More Information
Set Membership: |
|

Sec61g
Genomic Map