Assay Details
| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506720.2 | XP_003506768.1 | 3 - 4 | 548 | 54 | |
| GenBank mRNA | JI862495.1 | - | 3 - 4 | 535 | 54 | |
| JP048674.1 | - | 3 - 4 | 535 | 54 | ||
More Information
Set Membership: |
|

Timm22
Genomic Map