Assay Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507268.2 | XP_003507316.1 | 10 - 11 | 1421 | 58 | |
| XM_007647838.1 | XP_007646028.1 | 10 - 11 | 1411 | 58 | ||
| GenBank mRNA | JI876569.1 | - | 9 - 10 | 1393 | 58 | |
| JP042791.1 | - | 9 - 10 | 1393 | 58 | ||
More Information
Set Membership: |
|

Pafah1b1
Genomic Map