Assay Details
| Gene Symbol: Ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645258.1 | XP_007643448.1 | 4 - 5 | 354 | 59 | |
| GenBank mRNA | JP043095.1 | - | 4 - 5 | 354 | 59 | |
More Information
Set Membership: |
|

Ube2d4
Genomic Map