Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514038.2 | XP_003514086.1 | 3 - 4 | 606 | 72 | |
| GenBank mRNA | JI872168.1 | - | 3 - 4 | 536 | 72 | |
| JP044131.1 | - | 3 - 4 | 536 | 72 | ||
More Information
Set Membership: |
|

Tmem248
Genomic Map