Assay Details
| Gene Symbol: Il25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646594.1 | XP_007644784.1 | 4 - 5 | 1277 | 97 | |
| XM_007646595.1 | XP_007644785.1 | 4 - 5 | 1366 | 97 | ||
| XM_007646596.1 | XP_007644786.1 | 4 - 5 | 661 | 97 | ||
| XM_007646597.1 | XP_007644787.1 | 4 - 5 | 1341 | 97 | ||
| XM_007646598.1 | XP_007644788.1 | 5 - 6 | 1528 | 97 | ||
| XM_007646599.1 | XP_007644789.1 | 3 - 4 | 497 | 97 | ||
| XM_007646600.1 | XP_007644790.1 | 3 - 4 | 410 | 97 | ||
More Information
Set Membership: |
|

Il25
Genomic Map