Assay Details
| Gene Symbol: Psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497906.2 | XP_003497954.1 | 3 - 4 | 180 | 73 | |
| GenBank mRNA | JI896530.1 | - | 3 - 4 | 188 | 73 | |
| JP063369.1 | - | 3 - 4 | 188 | 73 | ||
More Information
Set Membership: |
|

Psma4
Genomic Map