Assay Details
| Gene Symbol: Slpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504574.2 | XP_003504622.1 | 3 - 4 | 681 | 75 | |
| GenBank mRNA | JI886786.1 | - | 3 - 4 | 781 | 75 | |
More Information
Set Membership: |
|

Slpi
Genomic Map