Assay Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506391.2 | XP_003506439.1 | 4 - 5 | 2379 | 70 | |
| XM_007646900.1 | XP_007645090.1 | 4 - 5 | 2386 | 70 | ||
| XM_007646901.1 | XP_007645091.1 | 3 - 4 | 2289 | 70 | ||
| GenBank mRNA | JI867085.1 | - | 3 - 4 | 2318 | 70 | |
| JP053409.1 | - | 3 - 4 | 2317 | 70 | ||
More Information
Set Membership: |
|

Lrrc8b
Genomic Map